Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Fluorbenzen ≥99%

Fluorbenzen ≥99%

Glass bottle for solids, 1.000 g
Catalog # A14384.0B
Flammable
Irritant
Fluorbenzen ≥99%
Catalog # A14384.0B
CAS Number:  462-06-6
Fluorbenzen ≥99%
Fluorbenzen ≥99%

Glass bottle for solids, 1.000 g

CAS Number:  462-06-6
Price
1.430,00 kr
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 462-06-6 Molecular Formula: C6H5F Formula Weight: 96.10 Flash Point: -15°(-5°F) Refractive Index: 1.4660 Odour: Aromatic Melting Point: -42° Boiling Point: 84-85° Beilstein Reference: 1236623 Merck Reference: 14,4170 UN No.: 2387 MDL No.: MFCD00000280

Synonyms:  Phenylfluoride
  • CAS Number:462-06-6
  • Formula:C₆H₅F
  • MDL Number:MFCD00000280
  • Molecular Weight:96.1 g/mol
  • Merck Index:14,4170
  • Boiling Pt:84…85 °C
  • Melting Pt:–42 °C
  • Density:1,025
  • Flash Pt:–15 °C (5 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:207-321-7
  • UN:2387
  • ADR:3,II
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.