5,5'-Dithiobis(2-nitrobenzosyre) ≥99%
Glass bottle for solids, 25 g
Purity: 99% CAS No.: 69-78-3 Molecular Formula: C14H8N2O8S2 Formula Weight: 396.36 Melting Point: ca 240° dec. Beilstein Reference: 1896821
: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
- :69-78-3
- :C₁₄H₈N₂O₈S₂
- :MFCD00007140
- :396.36 g/mol
- :∼ 240 °C (dec.)
- :Stue temperatur
- :200-714-4
- Pakning:Glass bottle for solids
- Pakn:25 g