5,5'-Dithiobis(2-nitrobenzosyre) ≥99%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
5,5'-Dithiobis(2-nitrobenzosyre) ≥99%
Katalog # A14331.06
Leverandør: Thermo Fisher Scientific
CAS nummer: 69-78-3
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 69-78-3 Molecular Formula: C14H8N2O8S2 Formula Weight: 396.36 Melting Point: ca 240° dec. Beilstein Reference: 1896821
Synonyms: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
- CAS Number:69-78-3
- Formula:C₁₄H₈N₂O₈S₂
- MDL Number:MFCD00007140
- Molecular Weight:396.36 g/mol
- Melting Pt:∼ 240 °C (dec.)
- Storage Temperature:Stue temperatur
- EINECS:200-714-4
Documents
Certificates
Search by Lot NumberALFAA14331.03_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



