Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
5,5'-Dithiobis(2-nitrobenzosyre) ≥99%

5,5'-Dithiobis(2-nitrobenzosyre) ≥99%

Glass bottle for solids, 1 g
Catalog # A14331.03
Irritant
5,5'-Dithiobis(2-nitrobenzosyre) ≥99%
Catalog # A14331.03
CAS Number:  69-78-3
5,5'-Dithiobis(2-nitrobenzosyre) ≥99%
5,5'-Dithiobis(2-nitrobenzosyre) ≥99%

Glass bottle for solids, 1 g

CAS Number:  69-78-3
Price
349,00 kr
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 99% CAS No.: 69-78-3 Molecular Formula: C14H8N2O8S2 Formula Weight: 396.36 Melting Point: ca 240° dec. Beilstein Reference: 1896821

Synonyms:  DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
  • CAS Number:69-78-3
  • Formula:C₁₄H₈N₂O₈S₂
  • MDL Number:MFCD00007140
  • Molecular Weight:396.36 g/mol
  • Melting Pt:∼ 240 °C (dec.)
  • Storage Temperature:Stue temperatur
  • EINECS:200-714-4
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.