Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methyl 4-hydroxybenzoat ≥99%

Methyl 4-hydroxybenzoat ≥99%

Plastic bottle for solids, 1.000 g
Katalog # A14289.0B
Irriterende
Leverandør:   Thermo Fisher Scientific
Methyl 4-hydroxybenzoat ≥99%
Katalog # A14289.0B
CAS nummer:  99-76-3
Methyl 4-hydroxybenzoat ≥99%
Methyl 4-hydroxybenzoat ≥99%

Plastic bottle for solids, 1.000 g

CAS nummer:  99-76-3
Price
1.820,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 99-76-3 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless pH: 5.8 Melting Point: 125-128° Boiling Point: ca 280° dec. Beilstein Reference: 509801 Merck Reference: 14,6107 MDL No.: MFCD00002352

Synonymer:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS nummer:99-76-3
  • Formel:HOC₆H₄CO₂CH₃
  • MDL nummer:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,6107
  • Kogende Pt:∼280 °C (dec.)
  • Smeltende Pt:125…128 °C
  • Tæthed:1,1208 g/cm³ (137 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-785-7
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    1.000 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.