Phenylboronsyre ≥98%
Glass bottle for solids, 10 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98+% CAS No.: 98-80-6 Molecular Formula: C6H7BO2 Formula Weight: 121.93 Melting Point: 214-219° Beilstein Reference: 970972 Merck Reference: 14,1068 MDL No.: MFCD00002103
Synonymer: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid
- CAS nummer:98-80-6
- Formel:C₆H₇BO₂
- MDL nummer:MFCD00002103
- Molecular Weight:121.93 g/mol
- Merck Index:14,1068
- Smeltende Pt:214…219 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-701-9
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g



