Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cyclopentanon ≥99%

Glass bottle, 500 mL
Katalog # A14222.AP
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Cyclopentanon ≥99%
Katalog # A14222.AP
CAS nummer:  120-92-3
Cyclopentanon ≥99%
Cyclopentanon ≥99%

Glass bottle, 500 mL

CAS nummer:  120-92-3
Price
678,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 120-92-3 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 30°(86°F) Refractive Index: 1.4370 Odour: Petrol-like Melting Point: -51° Boiling Point: 130-131° Beilstein Reference: 605573 Merck Reference: 14,2743 UN No.: 2245 MDL No.: MFCD00001409

  • CAS nummer:120-92-3
  • Formel:C₅H₈O
  • MDL nummer:MFCD00001409
  • Molecular Weight:84.12 g/mol
  • Merck Index:14,2743
  • Kogende Pt:130…131 °C
  • Smeltende Pt:–51 °C
  • Tæthed:0,949
  • Flash Pt:30 °C (86 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-435-9
  • FN:2245
  • ADR:3,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL
  • Cat. No.:
    A14222.AP

Specifikationer