Orders LinkContactUsLinkComponent
Cyclopentanon ≥99%
Glass bottle, 250 mL
  A14222.AK
Cyclopentanon ≥99%
  A14222.AK
 :  120-92-3
Cyclopentanon ≥99%
Cyclopentanon ≥99%
Glass bottle, 250 mL
 :  120-92-3
409,00 kr
1 * 250 mL
Purity: 99% CAS No.: 120-92-3 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 30°(86°F) Refractive Index: 1.4370 Odour: Petrol-like Melting Point: -51° Boiling Point: 130-131° Beilstein Reference: 605573 Merck Reference: 14,2743 UN No.: 2245 MDL No.: MFCD00001409
  •  :120-92-3
  •  :C₅H₈O
  •  :MFCD00001409
  •  :84.12 g/mol
  •  :14,2743
  •  :130…131 °C
  •  :–51 °C
  •  :0,949
  •  :30 °C (86 °F)
  •  :Stue temperatur
  •  :204-435-9
  •  :2245
  •  :3,III
  • Pakning:
    Glass bottle
  • Pakn:
    250 mL