Cyclopentanon ≥99%
Glass bottle, 250 mL
Purity: 99% CAS No.: 120-92-3 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 30°(86°F) Refractive Index: 1.4370 Odour: Petrol-like Melting Point: -51° Boiling Point: 130-131° Beilstein Reference: 605573 Merck Reference: 14,2743 UN No.: 2245 MDL No.: MFCD00001409
- :120-92-3
- :C₅H₈O
- :MFCD00001409
- :84.12 g/mol
- :14,2743
- :130…131 °C
- :–51 °C
- :0,949
- :30 °C (86 °F)
- :Stue temperatur
- :204-435-9
- :2245
- :3,III
- Pakning:Glass bottle
- Pakn:250 mL