2-Decanon ≥97%
Glass bottle for solids, 25 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 97% CAS No.: 693-54-9 Molecular Formula: CH3(CH2)7COCH3 Formula Weight: 156.27 Flash Point: 86°(187°F) Refractive Index: 1.4250 Melting Point: 3° Boiling Point: 210-212° Beilstein Reference: 1747463 MDL No.: MFCD00009571
Synonymer: Decan-2-one
- CAS nummer:693-54-9
- Formel:C₁₀H₂₀O
- MDL nummer:MFCD00009571
- Molecular Weight:156.27 g/mol
- Kogende Pt:210…212 °C
- Smeltende Pt:3 °C
- Tæthed:0,824
- Flash Pt:86 °C (187 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:211-752-6
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



