Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Decanon ≥97%

2-Decanon ≥97%

Glass bottle for solids, 5 g
Katalog # A14145.06
Leverandør:   Thermo Fisher Scientific
2-Decanon ≥97%
Katalog # A14145.06
CAS nummer:  693-54-9
2-Decanon ≥97%
2-Decanon ≥97%

Glass bottle for solids, 5 g

CAS nummer:  693-54-9
Price
338,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 693-54-9 Molecular Formula: CH3(CH2)7COCH3 Formula Weight: 156.27 Flash Point: 86°(187°F) Refractive Index: 1.4250 Melting Point: 3° Boiling Point: 210-212° Beilstein Reference: 1747463 MDL No.: MFCD00009571

Synonyms:  Decan-2-one
  • CAS Number:693-54-9
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00009571
  • Molecular Weight:156.27 g/mol
  • Boiling Pt:210…212 °C
  • Melting Pt:3 °C
  • Density:0,824
  • Flash Pt:86 °C (187 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:211-752-6
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.