Glycin ≥99%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS nummer:56-40-6
- Formel:NH₂CH₂COOH
- MDL nummer:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:14,4491
- Kogende Pt:181 °C (1013 hPa)
- Smeltende Pt:∼ 245 °C (dec.)
- Tæthed:1,595
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-272-2
Specifikationer
Specifikationstestresultater
- Appearance (Color):White
- Form:crystalline powder
- Assay (Non-aqueous acid-base Titration):98.5%
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



