Glycin ≥99%
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:14,4491
- Boiling Pt:181 °C (1013 hPa)
- Melting Pt:∼ 245 °C (dec.)
- Density:1,595
- Storage Temperature:Stue temperatur
- EINECS:200-272-2
Documents
Specifications
Specification Test Results
- Appearance (Color):White
- Form:crystalline powder
- Assay (Non-aqueous acid-base Titration):98.5%
Specifications
- Pakning:Glass bottle for solids
- Pakn:2.500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



