Glycin ≥99%
Glass bottle for solids, 10.000 g
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :14,4491
- :181 °C (1013 hPa)
- :∼ 245 °C (dec.)
- :1,595
- :Stue temperatur
- :200-272-2
- Appearance (Color):White
- Form:crystalline powder
- Assay (Non-aqueous acid-base Titration):98.5%
- Pakning:Glass bottle for solids
- Pakn:10.000 g



