Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Hexansyre ≥98%

Hexansyre ≥98%

Glass bottle, 2.500 mL
Katalog # A13789.0F
Giftig
Ætsende
Leverandør:   Thermo Fisher Scientific
Hexansyre ≥98%
Katalog # A13789.0F
CAS nummer:  142-62-1
Hexansyre ≥98%
Hexansyre ≥98%

Glass bottle, 2.500 mL

CAS nummer:  142-62-1
Price
kr1,840.00
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 98+% CAS No.: 142-62-1 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 104°(219°F) Physical Form: Clear liquid Density: 0.929 Refractive Index: 1.4165 Appearance: Pale yellow Melting Point: -4° Boiling Point: 202-203° Beilstein Reference: 773837 Merck Reference: 14,1759 UN No.: 2829 MDL No.: MFCD00004421

Synonymer:  Hexanoic acid
  • CAS nummer:142-62-1
  • Formel:C₆H₁₂O₂
  • MDL nummer:MFCD00004421
  • Molecular Weight:116.16 g/mol
  • Merck Index:14,1759
  • Kogende Pt:202…203 °C
  • Smeltende Pt:–4 °C
  • Tæthed:0,929
  • Flash Pt:104 °C (219 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:205-550-7
  • FN:2829
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer