Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Geraniol ≥97%

Geraniol ≥97%

Glass bottle for solids, 250 g
Katalog # A13736.30
Irritant
Leverandør:   Thermo Fisher Scientific
Geraniol ≥97%
Katalog # A13736.30
CAS nummer:  106-24-1
Geraniol ≥97%
Geraniol ≥97%

Glass bottle for solids, 250 g

CAS nummer:  106-24-1
Price
1.550,00 kr
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 97% CAS No.: 106-24-1 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 101°(213°F) Refractive Index: 1.4760 Odour: Floral Melting Point: -15° Boiling Point: 114-115°/12mm Beilstein Reference: 1722456 Merck Reference: 14,4403 MDL No.: MFCD00002917

Synonyms:  (E)-3,7-Dimethylocta-2,6-dien-1-ol
  • CAS Number:106-24-1
  • Formula:C₁₀H₁₈O
  • MDL Number:MFCD00002917
  • Molecular Weight:154.25 g/mol
  • Merck Index:14,4403
  • Boiling Pt:114…115 °C (12 mmHg)
  • Melting Pt:–15 °C
  • Density:0,884
  • Flash Pt:101 °C (213 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:203-377-1
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.