Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Geraniol ≥97%

Geraniol ≥97%

Glass bottle for solids, 1.000 g
Katalog # A13736.0B
Irriterende
Leverandør:   Thermo Fisher Scientific
Geraniol ≥97%
Katalog # A13736.0B
CAS nummer:  106-24-1
Geraniol ≥97%
Geraniol ≥97%

Glass bottle for solids, 1.000 g

CAS nummer:  106-24-1
Price
4.620,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

Purity: 97% CAS No.: 106-24-1 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 101°(213°F) Refractive Index: 1.4760 Odour: Floral Melting Point: -15° Boiling Point: 114-115°/12mm Beilstein Reference: 1722456 Merck Reference: 14,4403 MDL No.: MFCD00002917

Synonymer:  (E)-3,7-Dimethylocta-2,6-dien-1-ol
  • CAS nummer:106-24-1
  • Formel:C₁₀H₁₈O
  • MDL nummer:MFCD00002917
  • Molecular Weight:154.25 g/mol
  • Merck Index:14,4403
  • Kogende Pt:114…115 °C (12 mmHg)
  • Smeltende Pt:–15 °C
  • Tæthed:0,884
  • Flash Pt:101 °C (213 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-377-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.