(S)-(-)-Æblesyre ≥97%
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
- CAS nummer:97-67-6
- Formel:C₄H₆O₅
- MDL nummer:MFCD00064213
- Molecular Weight:134.09 g/mol
- Merck Index:14,5707
- Kogende Pt:322 °C (1013 hPa)
- Smeltende Pt:100…106 °C
- Tæthed:1,60
- Flash Pt:220 °C (428 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-601-5
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



