Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(S)-(-)-Æblesyre ≥97%

(S)-(-)-Æblesyre ≥97%

Glass bottle for solids, 250 g
Katalog # A13702.30
Irriterende
Leverandør:   Thermo Fisher Scientific
(S)-(-)-Æblesyre ≥97%
Katalog # A13702.30
CAS nummer:  97-67-6
(S)-(-)-Æblesyre ≥97%
(S)-(-)-Æblesyre ≥97%

Glass bottle for solids, 250 g

CAS nummer:  97-67-6
Price
1.690,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
  • CAS nummer:97-67-6
  • Formel:C₄H₆O₅
  • MDL nummer:MFCD00064213
  • Molecular Weight:134.09 g/mol
  • Merck Index:14,5707
  • Kogende Pt:322 °C (1013 hPa)
  • Smeltende Pt:100…106 °C
  • Tæthed:1,60
  • Flash Pt:220 °C (428 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-601-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.