Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
p-Cresol ≥99%

p-Cresol ≥99%

Glass bottle for solids, 250 g
Katalog # A13531.30
Giftig
Ætsende
Leverandør:   Thermo Fisher Scientific
p-Cresol ≥99%
Katalog # A13531.30
CAS nummer:  106-44-5
p-Cresol ≥99%
p-Cresol ≥99%

Glass bottle for solids, 250 g

CAS nummer:  106-44-5
Price
726,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 106-44-5 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 89°(192°F) Odour: Phenol-like Boiling Point: 201-202° Beilstein Reference: 1305151 Merck Reference: 14,2579 UN No.: 3455 MDL No.: MFCD00002376

Synonymer:  p-Methylphenol
  • CAS nummer:106-44-5
  • Formel:C₇H₈O
  • MDL nummer:MFCD00002376
  • Molecular Weight:108.14 g/mol
  • Merck Index:14,2579
  • Kogende Pt:201…202 °C
  • Smeltende Pt:32…34 °C
  • Tæthed:1,034
  • Flash Pt:89 °C (192 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-398-6
  • FN:3455
  • ADR:6.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer