Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
p-Cresol ≥99%

p-Cresol ≥99%

Glass bottle for solids, 100 g
Catalog # A13531.22
Toxic
Corrosive
p-Cresol ≥99%
Catalog # A13531.22
CAS Number:  106-44-5
p-Cresol ≥99%
p-Cresol ≥99%

Glass bottle for solids, 100 g

CAS Number:  106-44-5
Price
590,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 106-44-5 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 89°(192°F) Odour: Phenol-like Boiling Point: 201-202° Beilstein Reference: 1305151 Merck Reference: 14,2579 UN No.: 3455 MDL No.: MFCD00002376

Synonyms:  p-Methylphenol
  • CAS Number:106-44-5
  • Formula:C₇H₈O
  • MDL Number:MFCD00002376
  • Molecular Weight:108.14 g/mol
  • Merck Index:14,2579
  • Boiling Pt:201…202 °C
  • Melting Pt:32…34 °C
  • Density:1,034
  • Flash Pt:89 °C (192 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:203-398-6
  • UN:3455
  • ADR:6.1,II
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.