Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
p-Cresol ≥99%

p-Cresol ≥99%

Glass bottle for solids, 5.000 g
Katalog # A13531.0I
Giftig
Ætsende
Leverandør:   Thermo Fisher Scientific
p-Cresol ≥99%
Katalog # A13531.0I
CAS nummer:  106-44-5
p-Cresol ≥99%
p-Cresol ≥99%

Glass bottle for solids, 5.000 g

CAS nummer:  106-44-5
Price
6.800,00 kr
Log ind for tilgængelighed
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 106-44-5 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 89°(192°F) Odour: Phenol-like Boiling Point: 201-202° Beilstein Reference: 1305151 Merck Reference: 14,2579 UN No.: 3455 MDL No.: MFCD00002376

Synonymer:  p-Methylphenol
  • CAS nummer:106-44-5
  • Formel:C₇H₈O
  • MDL nummer:MFCD00002376
  • Molecular Weight:108.14 g/mol
  • Merck Index:14,2579
  • Kogende Pt:201…202 °C
  • Smeltende Pt:32…34 °C
  • Tæthed:1,034
  • Flash Pt:89 °C (192 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-398-6
  • FN:3455
  • ADR:6.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5.000 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.