Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Octan ≥98%

n-Octan ≥98%

Glass bottle, 500 mL
Katalog # A13181.AP
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
n-Octan ≥98%
Katalog # A13181.AP
CAS nummer:  111-65-9
n-Octan ≥98%
n-Octan ≥98%

Glass bottle, 500 mL

CAS nummer:  111-65-9
Price
1.100,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Purity: 98+% CAS No.: 111-65-9 Molecular Formula: CH3(CH2)6CH3 Formula Weight: 114.23 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.703 Refractive Index: 1.3980 Odour: Nearly Odourless Melting Point: -57° Boiling Point: 125-126° Solubility: Soluble in ether, Slightly soluble in alcohol Beilstein Reference: 1696875 Merck Reference: 14,6749 UN No.: 1262 MDL No.: MFCD00009556

  • CAS nummer:111-65-9
  • Formel:H₃C(CH₂)₆CH₃
  • MDL nummer:MFCD00009556
  • Molecular Weight:114.23 g/mol
  • Merck Index:14,6749
  • Kogende Pt:125…126 °C
  • Smeltende Pt:–57 °C
  • Tæthed:0,703
  • Flash Pt:15 °C (59 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-892-1
  • FN:1262
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer