Orders LinkContactUsLinkComponent
n-Octan ≥98%
n-Octan ≥98%
Glass bottle, 100 mL
  A13181.AE
n-Octan ≥98%
  A13181.AE
 :  111-65-9
n-Octan ≥98%
n-Octan ≥98%
Glass bottle, 100 mL
 :  111-65-9
415,00 kr
1 * 100 mL
Purity: 98+% CAS No.: 111-65-9 Molecular Formula: CH3(CH2)6CH3 Formula Weight: 114.23 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.703 Refractive Index: 1.3980 Odour: Nearly Odourless Melting Point: -57° Boiling Point: 125-126° Solubility: Soluble in ether, Slightly soluble in alcohol Beilstein Reference: 1696875 Merck Reference: 14,6749 UN No.: 1262 MDL No.: MFCD00009556
  •  :111-65-9
  •  :H₃C(CH₂)₆CH₃
  •  :MFCD00009556
  •  :114.23 g/mol
  •  :14,6749
  •  :125…126 °C
  •  :–57 °C
  •  :0,703
  •  :15 °C (59 °F)
  •  :Stue temperatur
  •  :203-892-1
  •  :1262
  •  :3,II
  • Pakning:
    Glass bottle
  • Pakn:
    100 mL