n-Octan ≥98%
Glass bottle, 100 mL
Purity: 98+% CAS No.: 111-65-9 Molecular Formula: CH3(CH2)6CH3 Formula Weight: 114.23 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.703 Refractive Index: 1.3980 Odour: Nearly Odourless Melting Point: -57° Boiling Point: 125-126° Solubility: Soluble in ether, Slightly soluble in alcohol Beilstein Reference: 1696875 Merck Reference: 14,6749 UN No.: 1262 MDL No.: MFCD00009556
- :111-65-9
- :H₃C(CH₂)₆CH₃
- :MFCD00009556
- :114.23 g/mol
- :14,6749
- :125…126 °C
- :–57 °C
- :0,703
- :15 °C (59 °F)
- :Stue temperatur
- :203-892-1
- :1262
- :3,II
- Pakning:Glass bottle
- Pakn:100 mL