Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Octan ≥98%

n-Octan ≥98%

Glass bottle, 2.500 mL
Katalog # A13181.0F
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
n-Octan ≥98%
Katalog # A13181.0F
CAS nummer:  111-65-9
n-Octan ≥98%
n-Octan ≥98%

Glass bottle, 2.500 mL

CAS nummer:  111-65-9
Price
3.720,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 98+% CAS No.: 111-65-9 Molecular Formula: CH3(CH2)6CH3 Formula Weight: 114.23 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.703 Refractive Index: 1.3980 Odour: Nearly Odourless Melting Point: -57° Boiling Point: 125-126° Solubility: Soluble in ether, Slightly soluble in alcohol Beilstein Reference: 1696875 Merck Reference: 14,6749 UN No.: 1262 MDL No.: MFCD00009556

  • CAS nummer:111-65-9
  • Formel:H₃C(CH₂)₆CH₃
  • MDL nummer:MFCD00009556
  • Molecular Weight:114.23 g/mol
  • Merck Index:14,6749
  • Kogende Pt:125…126 °C
  • Smeltende Pt:–57 °C
  • Tæthed:0,703
  • Flash Pt:15 °C (59 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-892-1
  • FN:1262
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.