Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Hydroxybenzylalkohol ≥99%

4-Hydroxybenzylalkohol ≥99%

Glass bottle for solids, 100 g
Katalog # A13132.22
Irriterende
Leverandør:   Thermo Fisher Scientific
4-Hydroxybenzylalkohol ≥99%
Katalog # A13132.22
CAS nummer:  623-05-2
4-Hydroxybenzylalkohol ≥99%
4-Hydroxybenzylalkohol ≥99%

Glass bottle for solids, 100 g

CAS nummer:  623-05-2
Price
1.920,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 623-05-2 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 114-122° Boiling Point: 251-253° MDL No.: MFCD00004658

Synonymer:  (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol
  • CAS nummer:623-05-2
  • Formel:C₇H₈O₂
  • MDL nummer:MFCD00004658
  • Molecular Weight:124.14 g/mol
  • Kogende Pt:251…253 °C
  • Smeltende Pt:114…122 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:210-768-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.