Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Aminopyrazin 99%

2-Aminopyrazin 99%

Glass bottle for solids, 50 g
Katalog # A13052.18
Irriterende
Leverandør:   Thermo Fisher Scientific
2-Aminopyrazin 99%
Katalog # A13052.18
CAS nummer:  5049-61-6
2-Aminopyrazin 99%
2-Aminopyrazin 99%

Glass bottle for solids, 50 g

CAS nummer:  5049-61-6
Price
2.850,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 5049-61-6 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 118-121° Beilstein Reference: 107025

Synonymer:  Aminopyrazine, Pyrazin-2-amine
  • CAS nummer:5049-61-6
  • Formel:C₄H₅N₃
  • MDL nummer:MFCD00006137
  • Molecular Weight:95.1 g/mol
  • Kogende Pt:203 °C (1013 hPa)
  • Smeltende Pt:118…121 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:225-748-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.