Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-Aminopyrazin 99%

Glass bottle for solids, 5 g
Katalog # A13052.06
Irriterende
Leverandør:   Thermo Fisher Scientific
2-Aminopyrazin 99%
Katalog # A13052.06
CAS nummer:  5049-61-6
2-Aminopyrazin 99%
2-Aminopyrazin 99%

Glass bottle for solids, 5 g

CAS nummer:  5049-61-6
Price
508,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 5049-61-6 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 118-121° Beilstein Reference: 107025

Synonymer:  Aminopyrazine, Pyrazin-2-amine
  • CAS nummer:5049-61-6
  • Formel:C₄H₅N₃
  • MDL nummer:MFCD00006137
  • Molecular Weight:95.1 g/mol
  • Kogende Pt:203 °C (1013 hPa)
  • Smeltende Pt:118…121 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:225-748-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer