Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kanelalkohol ≥98%

Kanelalkohol ≥98%

Glass bottle for solids, 250 g
Katalog # A13025.30
Irriterende
Leverandør:   Thermo Fisher Scientific
Kanelalkohol ≥98%
Katalog # A13025.30
CAS nummer:  104-54-1
Kanelalkohol ≥98%
Kanelalkohol ≥98%

Glass bottle for solids, 250 g

CAS nummer:  104-54-1
Price
363,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 104-54-1 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 126°(258°F) Odour: Pleasant Boiling Point: 257-258° Beilstein Reference: 1903999 Merck Reference: 14,2302 MDL No.: MFCD00002921

Synonymer:  Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
  • CAS nummer:104-54-1
  • Formel:C₉H₁₀O
  • MDL nummer:MFCD00002921
  • Molecular Weight:134.18 g/mol
  • Merck Index:14,2302
  • Kogende Pt:257…258 °C
  • Smeltende Pt:32…35 °C
  • Tæthed:1,044
  • Flash Pt:126 °C (258 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-212-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.