Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kanelalkohol ≥98%

Kanelalkohol ≥98%

Glass bottle for solids, 1.000 g
Catalog # A13025.0B
Irritant
Kanelalkohol ≥98%
Catalog # A13025.0B
CAS Number:  104-54-1
Kanelalkohol ≥98%
Kanelalkohol ≥98%

Glass bottle for solids, 1.000 g

CAS Number:  104-54-1
Price
1.170,00 kr
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 98% CAS No.: 104-54-1 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 126°(258°F) Odour: Pleasant Boiling Point: 257-258° Beilstein Reference: 1903999 Merck Reference: 14,2302 MDL No.: MFCD00002921

Synonyms:  Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
  • CAS Number:104-54-1
  • Formula:C₉H₁₀O
  • MDL Number:MFCD00002921
  • Molecular Weight:134.18 g/mol
  • Merck Index:14,2302
  • Boiling Pt:257…258 °C
  • Melting Pt:32…35 °C
  • Density:1,044
  • Flash Pt:126 °C (258 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:203-212-3
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.