Natriumhydroxid ≥97%, pearls
Glass bottle for solids, 500 g
Purity: 97% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odour: Odourless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :14,8627
- :1390 °C
- :318 °C
- :2,13
- :Stue temperatur
- :215-185-5
- :1823
- :8,II
- :01-2119457892-27
- Pakning:Glass bottle for solids
- Pakn:500 g