Natriumhydroxid ≥97%, pearls
Plastic bottle for solids, 2.500 g
Leverandør: Thermo Fisher Scientific
Natriumhydroxid ≥97%, pearls
Katalog # B24414.0E
Leverandør: Thermo Fisher Scientific
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Purity: 97% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odour: Odourless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,8627
- Kogende Pt:1390 °C
- Smeltende Pt:318 °C
- Tæthed:2,13
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-185-5
- FN:1823
- ADR:8,II
- NÅ:01-2119457892-27
Specifikationer
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:2.500 g



