Orders LinkContactUsLinkComponent
Congo red indicator
Glass bottle for solids, 500 g
  B24310.36
Congo red indicator
  B24310.36
 :  573-58-0
Congo red indicator
Congo red indicator
Glass bottle for solids, 500 g
 :  573-58-0
4.610,00 kr
1 * 500 g
CAS No.: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Formula Weight: 696.67 Odour: Odourless Melting Point: 360° Beilstein Reference: 3894858 Merck Reference: 14,2498 MDL No.: MFCD00004028
 :  CI 22120, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalen-1-sulfonat)
  •  :573-58-0
  •  :C₃₂H₂₂N₆Na₂O₆S₂
  •  :MFCD00004028
  •  :696.67 g/mol
  •  :14,2498
  •  :>360 °C
  •  :Stue temperatur
  •  :209-358-4
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g