Congo red indicator
Glass bottle for solids, 500 g
CAS No.: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Formula Weight: 696.67 Odour: Odourless Melting Point: 360° Beilstein Reference: 3894858 Merck Reference: 14,2498 MDL No.: MFCD00004028
: CI 22120, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalen-1-sulfonat)
- :573-58-0
- :C₃₂H₂₂N₆Na₂O₆S₂
- :MFCD00004028
- :696.67 g/mol
- :14,2498
- :>360 °C
- :Stue temperatur
- :209-358-4
- Pakning:Glass bottle for solids
- Pakn:500 g