Orders LinkContactUsLinkComponent
Fuchsin (syre)
Glass bottle for solids, 500 g
Katalog # B22222.36
Irriterende
Leverandør:   Thermo Fisher Scientific
Fuchsin (syre)
Katalog # B22222.36
CAS nummer:  3244-88-0
Fuchsin (syre)
Fuchsin (syre)
Glass bottle for solids, 500 g
CAS nummer:  3244-88-0
Price
9.850,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
CAS No.: 3244-88-0 Molecular Formula: C20H17N3Na2O9S3 Formula Weight: 585.53 Beilstein Reference: 4111656 Merck Reference: 14,107 MDL No.: MFCD00013286
Synonymer:  CI 42685
  • CAS nummer:3244-88-0
  • Formel:C₂₀H₁₇N₃Na₂O₉S₃
  • MDL nummer:MFCD00013286
  • Molecular Weight:585.55 g/mol
  • Merck Index:14,107
  • Smeltende Pt:> 130 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:221-816-5
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g
Specifikationer