Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phenol rød natrium salt

Phenol rød natrium salt

Glass bottle for solids, 25 g
Katalog # B21948.14
Irriterende
Leverandør:   Thermo Fisher Scientific
Phenol rød natrium salt
Katalog # B21948.14
CAS nummer:  34487-61-1
Phenol rød natrium salt
Phenol rød natrium salt

Glass bottle for solids, 25 g

CAS nummer:  34487-61-1
Price
805,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

CAS No.: 34487-61-1 Molecular Formula: C19H13NaO5S Formula Weight: 376.37 Melting Point: 285° dec. Beilstein Reference: 6260026 Merck Reference: 14,7247 MDL No.: MFCD00066901

Synonymer:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
  • CAS nummer:34487-61-1
  • Formel:C₁₉H₁₃NaO₅S
  • MDL nummer:MFCD00066901
  • Molecular Weight:376.35 g/mol
  • Merck Index:14,7247
  • Smeltende Pt:285 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:252-057-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.