Phenol rød natrium salt
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
Phenol rød natrium salt
Katalog # B21948.06
Leverandør: Thermo Fisher Scientific
CAS nummer: 34487-61-1
Gå til:
Product Details & Documents
CAS No.: 34487-61-1 Molecular Formula: C19H13NaO5S Formula Weight: 376.37 Melting Point: 285° dec. Beilstein Reference: 6260026 Merck Reference: 14,7247 MDL No.: MFCD00066901
Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
- CAS nummer:34487-61-1
- Formel:C₁₉H₁₃NaO₅S
- MDL nummer:MFCD00066901
- Molecular Weight:376.35 g/mol
- Merck Index:14,7247
- Smeltende Pt:285 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:252-057-8
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



