Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phenol rød

Phenol rød

Glass bottle for solids, 50 g
Katalog # B21710.18
Irriterende
Leverandør:   Thermo Fisher Scientific
Phenol rød
Katalog # B21710.18
CAS nummer:  143-74-8
Phenol rød
Phenol rød

Glass bottle for solids, 50 g

CAS nummer:  143-74-8
Price
1.300,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Odour: Odourless Boiling Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552

Synonymer:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS nummer:143-74-8
  • Formel:C₁₉H₁₄O₅S
  • MDL nummer:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Merck Index:14,7247
  • Kogende Pt:>300 °C
  • Smeltende Pt:> 300 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:205-609-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer