Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

3-Hydroxybenzonitril ≥99%

Glass bottle for solids, 5 g
Katalog # B21702.06
Irriterende
Leverandør:   Thermo Fisher Scientific
3-Hydroxybenzonitril ≥99%
Katalog # B21702.06
CAS nummer:  873-62-1
3-Hydroxybenzonitril ≥99%
3-Hydroxybenzonitril ≥99%

Glass bottle for solids, 5 g

CAS nummer:  873-62-1
Price
496,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 873-62-1 Molecular Formula: C7H5NO Formula Weight: 119.12 Melting Point: 78-81° MDL No.: MFCD00002252

Synonymer:  m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
  • CAS nummer:873-62-1
  • Formel:C₇H₅NO
  • MDL nummer:MFCD00002252
  • Molecular Weight:119.12 g/mol
  • Kogende Pt:265 °C (1013 hPa)
  • Smeltende Pt:78…81 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:212-845-4
  • FN:3276
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer