Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
X-Gal ≥98%

X-Gal ≥98%

Glass bottle for solids, 50 mg
Katalog # B21034.MF
Leverandør:   Thermo Fisher Scientific
X-Gal ≥98%
Katalog # B21034.MF
CAS nummer:  7240-90-6
X-Gal ≥98%
X-Gal ≥98%

Glass bottle for solids, 50 mg

CAS nummer:  7240-90-6
Price
685,00 kr
Log ind for tilgængelighed
1 * 50 mg
Product Details & Documents

Purity: 98+% CAS No.: 7240-90-6 Molecular Formula: C14H15BrClNO6 Formula Weight: 408.64 Storage Temperature: 2°C to 8°C Melting Point: ca 236° dec. Beilstein Reference: 1402009 Optical Rotation: -62° (c=1 in 50% DMF) MDL No.: MFCD00005666

Synonymer:  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  • CAS nummer:7240-90-6
  • Formel:C₁₄H₁₅BrClNO₆
  • MDL nummer:MFCD00005666
  • Molecular Weight:408.63 g/mol
  • Smeltende Pt:∼ 236 °C (dec.)
  • Opbevaringstemperatur:Køleskab
  • EINECS:230-640-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 mg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.