Chloramphenicol ≥99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 56-75-7 Molecular Formula: C11H12Cl2N2O5 Formula Weight: 323.14 Physical Form: Crystalline powder Appearance: White to pale cream Melting Point: 149-152° Beilstein Reference: 2225532 Merck Reference: 14,2077 Optical Rotation: +19° (c=6 in ethanol) MDL No.: MFCD00078159
Inhibitor of translation on the 50S subunit at the peptidyltransferase step
Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
- CAS Number:56-75-7
- Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
- MDL Number:MFCD00078159
- Molecular Weight:323.13 g/mol
- Merck Index:14,2077
- Boiling Pt:645 °C (1013 hPa)
- Melting Pt:149…152 °C
- Density:1,49 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-287-4
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow
Product Literature
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:100 g



