Orders LinkContactUsLinkComponent
Palmitinsyre 95%
Glass bottle for solids, 10.000 g
Katalog # B20322.0C
Leverandør:   Thermo Fisher Scientific
Palmitinsyre 95%
Katalog # B20322.0C
CAS nummer:  57-10-3
Palmitinsyre 95%
Palmitinsyre 95%
Glass bottle for solids, 10.000 g
CAS nummer:  57-10-3
Price
4.040,00 kr
Log ind for tilgængelighed
1 * 10.000 g
Product Details & Documents
Purity: 95% CAS No.: 57-10-3 Molecular Formula: C16H32O2 Formula Weight: 256.43 Flash Point: 206°(402°F) Refractive Index: 1.4273 Odour: Odourless Melting Point: 62-65° Boiling Point: 215°/15mm Beilstein Reference: 607489 Merck Reference: 14,6996 MDL No.: MFCD00002747
Synonymer:  Hexadecanoic acid, Hexadecylic acid
  • CAS nummer:57-10-3
  • Formel:C₁₆H₃₂O₂
  • MDL nummer:MFCD00002747
  • Molecular Weight:256.43 g/mol
  • Merck Index:14,6996
  • Kogende Pt:215 °C (15 mmHg)
  • Smeltende Pt:62…65 °C
  • Tæthed:0,853
  • Flash Pt:206 °C (402 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-312-9
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    10.000 g
Specifikationer