Orders LinkContactUsLinkComponent
Anisol ≥99%
Glass bottle for solids, 500 g
  A12997.36
Anisol ≥99%
  A12997.36
 :  100-66-3
Anisol ≥99%
Anisol ≥99%
Glass bottle for solids, 500 g
 :  100-66-3
505,00 kr
1 * 500 g
Purity: 99% CAS No.: 100-66-3 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 43°(109°F) Refractive Index: 1.5170 Odour: Ether-like Melting Point: -37° Boiling Point: 153-154° Beilstein Reference: 506892 Merck Reference: 14,669 UN No.: 2222 MDL No.: MFCD00008354
 :  Methyl phenyl ether, Methoxybenzene
  •  :100-66-3
  •  :C₇H₈O
  •  :MFCD00008354
  •  :108.14 g/mol
  •  :14,669
  •  :153…154 °C
  •  :–37 °C
  •  :0,994
  •  :43 °C (109 °F)
  •  :Stue temperatur
  •  :202-876-1
  •  :2222
  •  :3,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g