Anisol ≥99%
Glass bottle for solids, 2.500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 100-66-3 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 43°(109°F) Refractive Index: 1.5170 Odour: Ether-like Melting Point: -37° Boiling Point: 153-154° Beilstein Reference: 506892 Merck Reference: 14,669 UN No.: 2222 MDL No.: MFCD00008354
Synonymer: Methyl phenyl ether, Methoxybenzene
- CAS nummer:100-66-3
- Formel:C₇H₈O
- MDL nummer:MFCD00008354
- Molecular Weight:108.14 g/mol
- Merck Index:14,669
- Kogende Pt:153…154 °C
- Smeltende Pt:–37 °C
- Tæthed:0,994
- Flash Pt:43 °C (109 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-876-1
- FN:2222
- ADR:3,III
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:2.500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



