Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(+)-Valin ≥99%

L(+)-Valin ≥99%

Glass bottle for solids, 500 g
Katalog # A12720.36
Leverandør:   Thermo Fisher Scientific
L(+)-Valin ≥99%
Katalog # A12720.36
CAS nummer:  72-18-4
L(+)-Valin ≥99%
L(+)-Valin ≥99%

Glass bottle for solids, 500 g

CAS nummer:  72-18-4
Price
4.680,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 72-18-4 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Odour: Odourless Melting Point: ca 315° Beilstein Reference: 1721136 Merck Reference: 14,9909 Optical Rotation: +27° (c=5 in 5N HCl) MDL No.: MFCD00064220

Synonymer:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS nummer:72-18-4
  • Formel:C₅H₁₁NO₂
  • MDL nummer:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Merck Index:14,9909
  • Kogende Pt:217 °C (1013 hPa)
  • Smeltende Pt:∼ 315 °C
  • Tæthed:1,23
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-773-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.