Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cyclobutanol 96%

Glass bottle for solids, 1 g
Katalog # A12430.03
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Cyclobutanol 96%
Katalog # A12430.03
CAS nummer:  2919-23-5
Cyclobutanol 96%
Cyclobutanol 96%

Glass bottle for solids, 1 g

CAS nummer:  2919-23-5
Price
1.690,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Purity: 96% CAS No.: 2919-23-5 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: 21°(69°F) Refractive Index: 1.4360 Boiling Point: 123-124° Beilstein Reference: 2035937 UN No.: 1120 MDL No.: MFCD00001318

  • CAS nummer:2919-23-5
  • Formel:C₄H₈O
  • MDL nummer:MFCD00001318
  • Molecular Weight:72.11 g/mol
  • Kogende Pt:123…124 °C
  • Tæthed:0,925
  • Flash Pt:21 °C (69 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:220-858-1
  • FN:1120
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer