Orders LinkContactUsLinkComponent
L(+)-Leucin ≥99%
Glass bottle for solids, 500 g
  A12311.36
L(+)-Leucin ≥99%
  A12311.36
 :  61-90-5
L(+)-Leucin ≥99%
L(+)-Leucin ≥99%
Glass bottle for solids, 500 g
 :  61-90-5
2.590,00 kr
1 * 500 g
Purity: 99% CAS No.: 61-90-5 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Physical Form: Cryst powder Density: 1.293 Appearance: White Melting Point: 300° Beilstein Reference: 1721722 Merck Reference: 14,5451 Optical Rotation: +15° (c=2 in 5N HCl) MDL No.: MFCD00002617
 :  (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
  •  :61-90-5
  •  :C₆H₁₃NO₂
  •  :MFCD00002617
  •  :131.17 g/mol
  •  :14,5451
  •  :>300 °C
  •  :1,293
  •  :Stue temperatur
  •  :200-522-0
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g