Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Jod ≥99%

Jod ≥99%

Glass bottle for solids, 250 g
Catalog # A12278.30
Health
Irritant
Environment
Jod ≥99%
Catalog # A12278.30
CAS Number:  7553-56-2
Jod ≥99%
Jod ≥99%

Glass bottle for solids, 250 g

CAS Number:  7553-56-2
Price
1.130,00 kr
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99+% CAS No.: 7553-56-2 Molecular Formula: I2 Formula Weight: 253.81 Flash Point: None Physical Form: Cryst solid Density: 4.930 Appearance: Grey to purple Odour: Irritating Melting Point: 183-186° subl. Beilstein Reference: 969212 Merck Reference: 14,5014 UN No.: 3495 MDL No.: MFCD00011355

  • CAS Number:7553-56-2
  • Formula:I₂
  • MDL Number:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Merck Index:14,5014
  • Boiling Pt:184,3 °C (1013 hPa)
  • Melting Pt:114 °C
  • Density:4,9 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:231-442-4
  • UN:3495
  • ADR:8,III
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifications