Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Jod ≥99%

Jod ≥99%

Glass bottle for solids, 50 g
Katalog # A12278.18
Sundhed
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
Jod ≥99%
Katalog # A12278.18
CAS nummer:  7553-56-2
Jod ≥99%
Jod ≥99%

Glass bottle for solids, 50 g

CAS nummer:  7553-56-2
Price
485,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Purity: 99+% CAS No.: 7553-56-2 Molecular Formula: I2 Formula Weight: 253.81 Flash Point: None Physical Form: Cryst solid Density: 4.930 Appearance: Grey to purple Odour: Irritating Melting Point: 183-186° subl. Beilstein Reference: 969212 Merck Reference: 14,5014 UN No.: 3495 MDL No.: MFCD00011355

  • CAS nummer:7553-56-2
  • Formel:I₂
  • MDL nummer:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Merck Index:14,5014
  • Kogende Pt:184,3 °C (1013 hPa)
  • Smeltende Pt:114 °C
  • Tæthed:4,9 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-442-4
  • FN:3495
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.