Jod ≥99%
Glass bottle for solids, 1.000 g
Purity: 99+% CAS No.: 7553-56-2 Molecular Formula: I2 Formula Weight: 253.81 Flash Point: None Physical Form: Cryst solid Density: 4.930 Appearance: Grey to purple Odour: Irritating Melting Point: 183-186° subl. Beilstein Reference: 969212 Merck Reference: 14,5014 UN No.: 3495 MDL No.: MFCD00011355
- :7553-56-2
- :I₂
- :MFCD00011355
- :253.81 g/mol
- :14,5014
- :184,3 °C (1013 hPa)
- :114 °C
- :4,9 g/cm³ (20 °C)
- :Stue temperatur
- :231-442-4
- :3495
- :8,III
- Pakning:Glass bottle for solids
- Pakn:1.000 g