Stearinsyre ≥98%
Glass bottle for solids, 25 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98% CAS No.: 57-11-4 Molecular Formula: CH3(CH2)16CO2H Formula Weight: 284.48 Flash Point: 196°(384°F) Physical Form: Flakes or powder Density: 0.941 Appearance: White Odour: Odourless Melting Point: 69-72° Boiling Point: 232°/15mm Beilstein Reference: 608585 Merck Reference: 14,8804 MDL No.: MFCD00002752
A saturated fatty acid
Synonymer: Octadecanoic acid, Octadecylic acid
- CAS nummer:57-11-4
- Formel:C₁₈H₃₆O₂
- MDL nummer:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:14,8804
- Kogende Pt:232 °C (15 mmHg)
- Smeltende Pt:69…72 °C
- Tæthed:0,941
- Flash Pt:196 °C (384 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-313-4
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



