Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Formaldehyd dimethyl acetal ≥98%

Formaldehyd dimethyl acetal ≥98%

Glass bottle, 500 mL
Katalog # A12055.AP
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Formaldehyd dimethyl acetal ≥98%
Katalog # A12055.AP
CAS nummer:  109-87-5
Formaldehyd dimethyl acetal ≥98%
Formaldehyd dimethyl acetal ≥98%

Glass bottle, 500 mL

CAS nummer:  109-87-5
Price
430,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Purity: 98% CAS No.: 109-87-5 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: -17°(1°F) Refractive Index: 1.3545 Melting Point: -105° Boiling Point: 41-42° Beilstein Reference: 1697025 Merck Reference: 14,6012 UN No.: 1234 MDL No.: MFCD00008495

Synonymer:  Formal, Methylal, Dimethoxymethane
  • CAS nummer:109-87-5
  • Formel:CH₂(OCH₃)₂
  • MDL nummer:MFCD00008495
  • Molecular Weight:76.1 g/mol
  • Merck Index:14,6012
  • Kogende Pt:41…42 °C
  • Smeltende Pt:–105 °C
  • Tæthed:0,863
  • Flash Pt:–17 °C (1 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-714-2
  • FN:1234
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.